Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sbds
Entrez Gene ID
66711
Gene Name
Shwachman-Bodian-Diamond syndrome homolog (human)
Gene Aliases
4733401P19Rik, AI836084, CGI-97, SDS
Chromosome Location
Chr.5: 130245731 - 130255462 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023248.1 NP_075737.1 2 - 3 385 76
GenBank mRNA AK005079.1 - 2 - 3 362 76
AK014648.1 - 2 - 3 390 76
AK019228.1 - 2 - 3 372 76
AK168847.1 - 2 - 3 372 76
BC003849.1 - 2 - 3 385 76

View Full Product Details