The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081102.2 | NP_001074571.2 | 2 - 3 | 786 | 67 |
NM_001177884.1 | NP_001171355.1 | 2 - 3 | 786 | 67 | |
NM_175231.2 | NP_780440.2 | 2 - 3 | 786 | 67 | |
GenBank mRNA | AK038286.1 | - | 4 - 5 | 995 | 67 |
BC079668.1 | - | 2 - 3 | 702 | 67 | |
BC158094.1 | - | 3 - 4 | 860 | 67 | |
CF532498.1 | - | 2 - 3 | 323 | 67 |