Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Colq
Entrez Gene ID
382864
Gene Name
collagen-like tail subunit (single strand of homotrimer) of asymmetric acetylcholinesterase
Gene Aliases
A130034K24Rik
Chromosome Location
Chr.14: 31523082 - 31577863 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009937.2 NP_034067.2 1 - 2 241 61
GenBank mRNA AK134212.1 - 1 - 2 243 61
BC107386.2 - 1 - 2 175 61
BC107387.1 - 1 - 2 175 61

View Full Product Details