The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159577.1 | NP_001153049.1 | 7 - 8 | 1701 | 65 |
NM_001159578.1 | NP_001153050.1 | 8 - 9 | 1846 | 65 | |
NM_001159579.1 | NP_001153051.1 | 6 - 7 | 1477 | 65 | |
NM_001159580.1 | NP_001153052.1 | 6 - 7 | 1447 | 65 | |
NM_010727.4 | NP_034857.3 | 6 - 7 | 1477 | 65 | |
GenBank mRNA | AF034745.1 | - | 7 - 8 | 1701 | 65 |
AF034746.1 | - | 6 - 7 | 1405 | 65 | |
AK044127.1 | - | 6 - 7 | 1430 | 65 | |
AK138326.1 | - | 6 - 7 | 1448 | 65 | |
AK161194.1 | - | 8 - 9 | 1832 | 65 | |
BC139771.1 | - | 6 - 7 | 1405 | 65 | |
KJ418422.1 | - | 6 - 7 | 1175 | 65 |