Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spaca1
Entrez Gene ID
67652
Gene Name
sperm acrosome associated 1
Gene Aliases
1700124L11Rik, 4930540L03Rik
Chromosome Location
Chr.4: 34024872 - 34050067 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290443.1 NP_001277372.1 4 - 5 573 146
NM_026293.3 NP_080569.1 4 - 5 575 146
GenBank mRNA AK006187.1 - 4 - 5 576 146
AK007270.1 - 4 - 5 547 146
AK016011.1 - 4 - 5 568 146
BC048388.1 - 1 - 2 232 146
BC147079.1 - 4 - 5 572 146
BC147080.1 - 4 - 5 572 146
BY706285.1 - 4 - 5 574 146

View Full Product Details