Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Slc4a1ap
- Entrez Gene ID
-
20534
- Gene Name
- solute carrier family 4 (anion exchanger), member 1, adaptor protein
- Gene Aliases
-
kanadaptin
- Chromosome Location
-
Chr.5: 31526850 - 31554038 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK160250.1 |
- |
2 - 3 |
887 |
88 |
View Full Product Details