The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205070.1 | NP_001191999.1 | 2 - 3 | 326 | 73 |
GenBank mRNA | BC018244.1 | - | 2 - 3 | 155 | 73 |