Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rnf32
Entrez Gene ID
56874
Gene Name
ring finger protein 32
Gene Aliases
1700009J01Rik, 2700025B22Rik, 4930542N22Rik, Lmbr2
Chromosome Location
Chr.5: 29195170 - 29225526 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021470.6 NP_067445.3 1 - 2 92 124
GenBank mRNA AK005795.1 - 1 - 2 47 124
BY293373.1 - 1 - 2 91 124

View Full Product Details