The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012000.3 | NP_036130.1 | 1 - 2 | 585 | 64 |
GenBank mRNA | AF125307.1 | - | 1 - 2 | 75 | 64 |
AF125308.1 | - | 1 - 2 | 75 | 64 | |
AK049145.1 | - | 1 - 2 | 258 | 64 | |
AK080049.1 | - | 1 - 2 | 93 | 64 | |
AK088300.1 | - | 1 - 2 | 239 | 64 | |
AK089692.1 | - | 1 - 2 | 234 | 64 | |
AK128890.1 | - | 1 - 2 | 256 | 64 | |
AK164268.1 | - | 1 - 2 | 258 | 64 | |
AK166905.1 | - | 1 - 2 | 1010 | 64 | |
AK169698.1 | - | 1 - 2 | 239 | 64 | |
AK171446.1 | - | 1 - 2 | 267 | 64 | |
AK207099.1 | - | - | 29 | 64 | |
BC021625.1 | - | 1 - 2 | 91 | 64 |