Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Crtc2
- Entrez Gene ID
-
74343
- Gene Name
- CREB regulated transcription coactivator 2
- Gene Aliases
-
4632407F12Rik, Torc2, mTORC2
- Chromosome Location
-
Chr.3: 90254168 - 90264125 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK014553.1 |
- |
6 - 7 |
1908 |
128 |
View Full Product Details