The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_212452.1 | NP_997617.1 | 17 - 18 | 1965 | 146 |
GenBank mRNA | AK137655.1 | - | 15 - 16 | 4068 | 146 |
AY509975.1 | - | 17 - 18 | 1965 | 146 |