Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wfs1
Entrez Gene ID
22393
Gene Name
Wolfram syndrome 1 homolog (human)
Gene Aliases
AI481085, wolframin
Chromosome Location
Chr.5: 36966104 - 36988982 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011716.2 NP_035846.1 7 - 8 992 63
GenBank mRNA AF084482.1 - 7 - 8 925 63
AJ011971.1 - 7 - 8 919 63
AK145023.1 - 7 - 8 964 63
AK170180.1 - 7 - 8 947 63
BC046988.1 - 7 - 8 928 63
CX562372.1 - 5 - 6 622 63

View Full Product Details