Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ercc6
Entrez Gene ID
319955
Gene Name
excision repair cross-complementing rodent repair deficiency, complementation group 6
Gene Aliases
4732403I04, C130058G22Rik, CSB
Chromosome Location
Chr.14: 32513288 - 32580990 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081221.1 NP_001074690.1 3 - 4 633 74

View Full Product Details