The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205070.1 | NP_001191999.1 | 5 - 6 | 808 | 115 |
NM_001205071.1 | NP_001192000.1 | 4 - 5 | 696 | 115 | |
NM_001205072.1 | NP_001192001.1 | 4 - 5 | 665 | 115 | |
NM_001205073.1 | NP_001192002.1 | 4 - 5 | 662 | 115 | |
NM_025368.4 | NP_079644.1 | 4 - 5 | 693 | 115 | |
GenBank mRNA | AK002872.1 | - | 4 - 5 | 534 | 115 |
AK003525.1 | - | 4 - 5 | 618 | 115 | |
CA326008.1 | - | 4 - 5 | 607 | 115 |