Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ythdc1
Entrez Gene ID
231386
Gene Name
YTH domain containing 1
Gene Aliases
A730098D12Rik, C80342, mKIAA1966
Chromosome Location
Chr.5: 86803980 - 86836659 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177680.3 NP_808348.2 11 - 12 1685 78
GenBank mRNA AB221619.1 - 3 - 4 309 78
BC022697.1 - 4 - 5 478 78
BC048817.1 - 4 - 5 463 78

View Full Product Details