Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ythdc1
Entrez Gene ID
231386
Gene Name
YTH domain containing 1
Gene Aliases
A730098D12Rik, C80342, mKIAA1966
Chromosome Location
Chr.5: 86803980 - 86836659 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177680.3 NP_808348.2 15 - 16 1919 121
GenBank mRNA AB221619.1 - 7 - 8 519 121
AK080576.1 - 2 - 3 131 121
AK173312.1 - 11 - 12 3590 121
BC022697.1 - 8 - 9 688 121
BC037593.1 - 9 - 10 2146 121
BC044650.1 - 11 - 12 5196 121
BC048817.1 - 8 - 9 673 121

View Full Product Details