The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001650.1 | NP_001001650.1 | 3 - 4 | 254 | 98 |
GenBank mRNA | BC116378.1 | - | 3 - 4 | 254 | 98 |
BC116379.1 | - | 3 - 4 | 254 | 98 |