Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrp5
Entrez Gene ID
16973
Gene Name
low density lipoprotein receptor-related protein 5
Gene Aliases
BMND1, HBM, LR3, LRP7, OPPG, mKIAA4142
Chromosome Location
Chr.19: 3584825 - 3686564 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008513.3 NP_032539.2 10 - 11 2439 78
GenBank mRNA AF064984.1 - 10 - 11 2429 78
AF077847.1 - 10 - 11 2317 78
AK147068.1 - 10 - 11 2432 78
AK147231.1 - 11 - 12 2550 78
AK147330.1 - 11 - 12 2550 78
AK220186.1 - 10 - 11 2392 78
BC011374.1 - 10 - 11 2421 78
BC094610.1 - 10 - 11 2428 78

View Full Product Details