The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_054056.2 | NP_473397.1 | 6 - 7 | 1104 | 80 |
GenBank mRNA | AF377871.1 | - | 5 - 6 | 507 | 80 |
AK136412.1 | - | 1 - 2 | 69 | 80 | |
DQ363525.1 | - | 6 - 7 | 1104 | 80 | |
DQ449073.1 | - | 4 - 5 | 783 | 80 | |
DQ449074.1 | - | 5 - 6 | 915 | 80 | |
DV057979.1 | - | 1 - 2 | 80 |