Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdr82
Entrez Gene ID
77305
Gene Name
WD repeat domain containing 82
Gene Aliases
9430077D24Rik, CDW5/WDR82
Chromosome Location
Chr.9: 106170929 - 106191706 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029896.1 NP_084172.1 4 - 5 716 73
GenBank mRNA AK035109.1 - 4 - 5 718 73
AK037620.1 - 4 - 5 723 73
AK149692.1 - 4 - 5 717 73
AK159240.1 - 4 - 5 716 73
BC019115.1 - 3 - 4 242 73
BC031502.1 - 4 - 5 406 73
BC094419.1 - 4 - 5 679 73
BC157943.1 - - 639 73
BC157945.1 - - 640 73
BC158054.1 - - 640 73
EF011615.1 - 4 - 5 405 73

View Full Product Details