The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252466.1 | NP_001239395.1 | 10 - 11 | 1060 | 72 |
NM_001252467.1 | NP_001239396.1 | 7 - 8 | 700 | 72 | |
NM_027933.3 | NP_082209.1 | 9 - 10 | 859 | 72 | |
NR_045519.1 | 10 - 11 | 969 | 72 | ||
GenBank mRNA | AK011541.1 | - | 2 - 3 | 220 | 72 |
AK040481.1 | - | 11 - 12 | 1043 | 72 | |
AK133949.1 | - | 9 - 10 | 706 | 72 | |
AK154201.1 | - | 8 - 8 | 269 | ||
AK155301.1 | - | 9 - 10 | 728 | 72 | |
AK167731.1 | - | 9 - 10 | 709 | 72 | |
AK214950.1 | - | 3 - 4 | 291 | 72 | |
BC066005.1 | - | 9 - 10 | 700 | 72 | |
BC145892.1 | - | 9 - 10 | 723 | 72 | |
BC145894.1 | - | 9 - 10 | 723 | 72 |