Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cpne4
Entrez Gene ID
74020
Gene Name
copine IV
Gene Aliases
3632411M23Rik, 4933406O10Rik
Chromosome Location
Chr.9: 104547291 - 105034549 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028719.1 NP_082995.1 9 - 10 1184 76
GenBank mRNA AK043692.1 - 9 - 10 1197 76
AK191212.1 - 2 - 3 200 76
BC043087.1 - 9 - 10 1184 76
BC063081.1 - 9 - 10 1184 76

View Full Product Details