The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030133.3 | NP_084409.3 | 3 - 4 | 318 | 81 |
GenBank mRNA | AK021298.1 | - | 3 - 4 | 318 | 81 |