Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arrdc5
Entrez Gene ID
76920
Gene Name
arrestin domain containing 5
Gene Aliases
1700013E09Rik
Chromosome Location
Chr.17: 56294111 - 56300286 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029799.1 NP_084075.1 1 - 2 291 119
GenBank mRNA BC100496.1 - 1 - 2 291 119

View Full Product Details