Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fndc3c1
Entrez Gene ID
333564
Gene Name
fibronectin type III domain containing 3C1
Gene Aliases
Fndc3c, Gm784
Chromosome Location
Chr.X: 106420041 - 106485576 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007580.1 NP_001007581.1 22 - 23 3277 69
GenBank mRNA BC076625.1 - 22 - 23 3277 69
BC085176.1 - 22 - 23 3275 69
DQ192038.1 - 22 - 23 3322 69

View Full Product Details