The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198113.2 | NP_932781.1 | 13 - 14 | 1765 | 115 |
GenBank mRNA | AB099289.1 | - | 13 - 14 | 1763 | 115 |
AK148293.1 | - | 13 - 14 | 1758 | 115 | |
AK149880.1 | - | 13 - 14 | 1786 | 115 | |
AK164662.1 | - | 12 - 13 | 1464 | 115 | |
BC028922.1 | - | 13 - 14 | 1765 | 115 |