The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003910.2 | NP_001003910.2 | 7 - 8 | 2418 | 82 |
GenBank mRNA | AK133195.1 | - | 7 - 8 | 2359 | 82 |
AY651021.1 | - | 6 - 7 | 2058 | 82 |