Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tecr
Entrez Gene ID
106529
Gene Name
trans-2,3-enoyl-CoA reductase
Gene Aliases
2410016D23Rik, A230102P12Rik, AI173355, D17Ertd178e, Gpsn2, SC2
Chromosome Location
Chr.8: 83571698 - 83594491 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027179.1 NP_081455.1 1 - 2 149 60
NM_134118.5 NP_598879.1 1 - 2 149 60
GenBank mRNA AK002384.1 - 1 - 2 120 60
AK010513.1 - 1 - 2 85 60
AK010984.1 - 1 - 2 123 60
AK013558.1 - 1 - 2 105 60
AK020708.1 - 1 - 2 143 60
AK179321.1 - 1 - 2 58 60
BC019984.1 - 1 - 2 135 60
BC094046.1 - 1 - 2 87 60
BY278351.1 - 1 - 2 149 60

View Full Product Details