The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025551.3 | NP_079827.2 | 1 - 2 | 121 | 115 |
GenBank mRNA | AK003131.1 | - | 1 - 2 | 94 | 115 |
AK003322.1 | - | 1 - 2 | 94 | 115 | |
AK008876.1 | - | 1 - 2 | 94 | 115 | |
AK032774.1 | - | 1 - 2 | 95 | 115 | |
AK134765.1 | - | 1 - 2 | 94 | 115 | |
AK153240.1 | - | 1 - 2 | 94 | 115 | |
AK167943.1 | - | 1 - 2 | 87 | 115 | |
BC086922.1 | - | 1 - 2 | 126 | 115 |