Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hck
Entrez Gene ID
15162
Gene Name
hemopoietic cell kinase
Gene Aliases
AI849071, Bmk
Chromosome Location
Chr.2: 153108468 - 153151441 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172117.1 NP_001165588.1 2 - 3 401 74
NM_010407.4 NP_034537.2 2 - 3 401 74
GenBank mRNA AK150290.1 - 2 - 3 375 74
AK150709.1 - 2 - 3 384 74
AK152506.1 - 2 - 3 375 74
AK155975.1 - 2 - 3 74
AK159234.1 - 2 - 3 333 74
AK165315.1 - 2 - 3 366 74
BC010478.1 - 2 - 3 397 74
J03023.1 - 2 - 3 290 74
Y00487.1 - 2 - 3 296 74

View Full Product Details