The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199019.2 | NP_950184.2 | 1 - 1 | 2179 | 69 |
GenBank mRNA | AK077122.1 | - | 1 - 1 | 2181 | 69 |
AV279514.2 | - | 1 - 1 | 621 | 69 |