Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rprml
Entrez Gene ID
104582
Gene Name
reprimo-like
Gene Aliases
AW049332
Chromosome Location
Chr.11: 103649509 - 103650580 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033212.2 NP_001028384.1 1 - 1 922 118
GenBank mRNA AK141598.1 - 1 - 1 923 118
BC147064.1 - 1 - 1 897 118
BC147065.1 - 1 - 1 897 118

View Full Product Details