Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hook2
Entrez Gene ID
170833
Gene Name
hook homolog 2 (Drosophila)
Gene Aliases
A630054I03Rik
Chromosome Location
Chr.8: 84989501 - 85003364 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167991.1 NP_001161463.1 8 - 9 709 86
NM_133255.2 NP_573518.2 8 - 9 709 86
GenBank mRNA AK030599.1 - 8 - 9 702 86
AK042048.1 - 8 - 9 697 86
AK136069.1 - 8 - 9 703 86
AK145768.1 - 8 - 9 677 86
AK161612.1 - 8 - 9 662 86
AK166954.1 - 8 - 9 689 86
AK169073.1 - 8 - 9 677 86
BC019486.1 - 8 - 9 685 86
BC029967.1 - 8 - 9 650 86
BC055881.1 - 8 - 9 655 86
BC080744.1 - 8 - 9 699 86

View Full Product Details