Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nlgn2
Entrez Gene ID
216856
Gene Name
neuroligin 2
Gene Aliases
NL2
Chromosome Location
Chr.11: 69823123 - 69838377 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198862.2 NP_942562.2 4 - 5 1250 62
GenBank mRNA AK136883.1 - 5 - 6 1816 62
AK173159.1 - 4 - 5 1250 62
BE952199.1 - 4 - 5 454 62

View Full Product Details