The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277188.1 | NP_001264117.1 | 1 - 2 | 359 | 84 |
NR_102339.1 | 1 - 2 | 359 | 84 | ||
GenBank mRNA | AK039466.1 | - | 1 - 2 | 161 | 84 |
AK078364.1 | - | 1 - 2 | 123 | 84 | |
BC065114.1 | - | 1 - 2 | 359 | 84 | |
CF535597.1 | - | 1 - 2 | 359 | 84 |