Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dhx8
Entrez Gene ID
217207
Gene Name
DEAH (Asp-Glu-Ala-His) box polypeptide 8
Gene Aliases
Ddx8, mDEAH6, mKIAA4096
Chromosome Location
Chr.11: 101732956 - 101767358 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144831.2 NP_659080.2 6 - 7 1022 70
GenBank mRNA AK078552.1 - 6 - 7 961 70
AK170616.1 - 6 - 7 950 70
AK172600.1 - 6 - 7 967 70
AK220485.1 - 6 - 7 999 70
BC065064.1 - 6 - 7 951 70
BC132445.1 - 6 - 7 1006 70

View Full Product Details