Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fam83c
Entrez Gene ID
71405
Gene Name
family with sequence similarity 83, member C
Gene Aliases
5530400B04Rik
Chromosome Location
Chr.2: 155829183 - 155834854 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027788.2 NP_082064.2 2 - 3 777 84
GenBank mRNA BC113190.1 - 2 - 3 776 84
BC113797.1 - 2 - 3 776 84

View Full Product Details