The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178367.4 | NP_848144.3 | 6 - 7 | 1176 | 80 |
GenBank mRNA | AK035177.1 | - | 6 - 7 | 1176 | 80 |
BC027842.1 | - | 6 - 7 | 1173 | 80 | |
BC052172.1 | - | 6 - 7 | 1170 | 80 |