Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kctd2
Entrez Gene ID
70382
Gene Name
potassium channel tetramerisation domain containing 2
Gene Aliases
2310012I15Rik
Chromosome Location
Chr.11: 115420126 - 115431274 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183285.3 NP_899108.3 5 - 6 802 126
GenBank mRNA AA537633.1 - 5 - 6 456 126
BC095988.1 - 5 - 6 761 126

View Full Product Details