Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kif18b
Entrez Gene ID
70218
Gene Name
kinesin family member 18B
Gene Aliases
3000004C01Rik
Chromosome Location
Chr.11: 102905518 - 102925134 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_197959.2 NP_932063.2 2 - 3 470 52
GenBank mRNA AK156592.1 - 2 - 3 440 52
BC057614.1 - 2 - 3 438 52
BQ571943.1 - 2 - 3 433 52

View Full Product Details