Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arnt
Entrez Gene ID
11863
Gene Name
aryl hydrocarbon receptor nuclear translocator
Gene Aliases
D3Ertd557e, Drnt, ESTM42, Hif1b, W08714, bHLHe2, mKIAA4051
Chromosome Location
Chr.3: 95434368 - 95497240 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037737.2 NP_001032826.1 10 - 11 1103 70
NM_009709.4 NP_033839.2 9 - 10 1058 70
GenBank mRNA AK028546.1 - 9 - 10 1045 70
AK040475.1 - 10 - 11 1105 70
AK141362.1 - 10 - 11 974 70
AK145415.1 - 9 - 10 1013 70
AK152473.1 - 10 - 11 1085 70
AK153355.1 - 10 - 11 1085 70
AK220458.1 - 6 - 7 1234 70
BC012870.1 - 10 - 11 1065 70
U10325.1 - 10 - 11 967 70

View Full Product Details