Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyb5rl
Entrez Gene ID
230582
Gene Name
cytochrome b5 reductase-like
Gene Aliases
2810410C14Rik, mFLJ00377
Chromosome Location
Chr.4: 107066844 - 107087993 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY437136.1 - - 209 75
BC004750.1 - 1 - 2 218 75

View Full Product Details