Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Xpnpep1
Entrez Gene ID
170750
Gene Name
X-prolyl aminopeptidase (aminopeptidase P) 1, soluble
Gene Aliases
D230045I08Rik, sAMP
Chromosome Location
Chr.19: 52932227 - 53038654 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133216.3 NP_573479.3 18 - 19 1804 91
GenBank mRNA AK052094.1 - 10 - 11 946 91
AK080423.1 - 7 - 8 552 91
AK146033.1 - 18 - 19 1784 91
AK149676.1 - 18 - 19 1743 91
AK158597.1 - 16 - 17 1762 90
AK166715.1 - 18 - 19 1704 91
AK169024.1 - 17 - 18 1630 91
BC021534.1 - 9 - 10 739 91
BC065174.1 - 18 - 19 1692 91

View Full Product Details