Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tuft1
Entrez Gene ID
22156
Gene Name
tuftelin 1
Gene Aliases
-
Chromosome Location
Chr.3: 94612753 - 94658872 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293728.1 NP_001280657.1 10 - 11 1020 95
NM_011656.3 NP_035786.1 11 - 12 1095 95
GenBank mRNA AF047704.1 - 11 - 12 1029 95
AK132488.1 - 11 - 12 1027 95
AK171705.1 - 10 - 11 985 95
BC019213.1 - 11 - 12 1026 95

View Full Product Details