The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164583.1 | NP_001158055.1 | 9 - 10 | 1263 | 102 |
NM_001164584.1 | NP_001158056.1 | 9 - 10 | 1208 | 102 | |
NM_001164585.1 | NP_001158057.1 | 10 - 11 | 1273 | 102 | |
NM_198412.2 | NP_940804.1 | 10 - 11 | 1182 | 102 | |
GenBank mRNA | AK122293.1 | - | 9 - 10 | 1184 | 102 |
AK147570.1 | - | 10 - 11 | 1145 | 102 | |
AK147637.1 | - | 10 - 11 | 1145 | 102 | |
AK163657.1 | - | 9 - 10 | 1082 | 102 | |
BC035262.1 | - | 4 - 5 | 417 | 102 | |
BC060734.1 | - | 9 - 10 | 1263 | 102 | |
BC064460.1 | - | 10 - 11 | 1151 | 102 |