Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ankrd35
Entrez Gene ID
213121
Gene Name
ankyrin repeat domain 35
Gene Aliases
4732436F15Rik
Chromosome Location
Chr.3: 96670131 - 96691034 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081139.1 NP_001074608.1 4 - 5 601 75
GenBank mRNA AK028691.1 - 4 - 5 391 75
AK039772.1 - 4 - 5 356 75
AK046624.1 - 4 - 5 410 75
BC138276.1 - 4 - 5 403 75
BC145228.1 - 4 - 5 407 75
BC145750.1 - 4 - 5 403 75
BC157973.1 - 4 - 5 403 75

View Full Product Details