The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081139.1 | NP_001074608.1 | 6 - 7 | 740 | 73 |
GenBank mRNA | AK028691.1 | - | 6 - 7 | 530 | 73 |
AK039772.1 | - | 6 - 7 | 495 | 73 | |
AK046624.1 | - | 6 - 7 | 549 | 73 | |
BC138276.1 | - | 6 - 7 | 542 | 73 | |
BC145228.1 | - | 6 - 7 | 546 | 73 | |
BC145750.1 | - | 6 - 7 | 542 | 73 | |
BC157973.1 | - | 6 - 7 | 542 | 73 |