The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175140.4 | NP_780349.3 | 1 - 2 | 368 | 80 |
GenBank mRNA | AF308145.1 | - | 1 - 2 | 186 | 80 |
AK005217.1 | - | 1 - 2 | 110 | 80 | |
BC051124.1 | - | 1 - 2 | 333 | 80 | |
CF171795.1 | - | 1 - 2 | 323 | 80 |