Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tdrkh
Entrez Gene ID
72634
Gene Name
tudor and KH domain containing protein
Gene Aliases
2700091C21Rik, Tdrd2
Chromosome Location
Chr.3: 94413239 - 94432975 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028307.1 NP_082583.1 6 - 7 958 118
GenBank mRNA BC049363.1 - 6 - 7 958 118
BC057030.1 - 6 - 7 974 118

View Full Product Details