The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021429.3 | NP_067404.2 | 1 - 2 | 63 | 72 |
GenBank mRNA | AK040654.1 | - | 1 - 2 | 63 | 72 |
AK084802.1 | - | 1 - 2 | 56 | 72 | |
AK154696.1 | - | 1 - 2 | 59 | 72 | |
AK170838.1 | - | 1 - 2 | 50 | 72 | |
BC095977.1 | - | 1 - 2 | 60 | 72 |